Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand:  HMC Europe 50029
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10348082
  • € 699.60 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
Specifications
15.35 in.
22.45 gal.
HV 85 autoclave
290 mm
Stainless Steel
390 mm
85 L
11.41 in.
With collecting pan
390 x 290 mm