Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand:  HMC Europe 51010
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10793164
  • € 588.10 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
Specifications
13.3 in.
13.2/21.13 gal.
HG 50/80 autoclave
220 mm
Stainless Steel
345 mm
80 L (Total), 50 L (Nominal)
8.66 in.
Wire mesh basket with bottom
345 x 220 mm