missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5A Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76816H
This item is not returnable.
View return policy
Description
Kinesin 5A Polyclonal antibody specifically detects Kinesin 5A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| Kinesin 5A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| HRP | |
| D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10 | |
| Antibody was raised against a 20 amino acid synthetic peptide from near the carboxy terminus of human KIF5. The immunogen is located within amino acids 880 - 930 of KIF5. Amino Acid Squence: KDKRRYQQEVDRIKEAVRYK | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3798 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction