Learn More
Kinesin 5B Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32279AF647
This item is not returnable.
View return policy
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human samples. It is validated for Western Blot
Specifications
| Western Blot | |
| Alexa Fluor 647 | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| The immunogen this antibody was made to, maps to a region between residue 913 to 963 of human Kinesin Family Member 5B using the numbering given in entry NP_004512.1 (GeneID 3799). | |
| Primary | |
| IgG |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| Purified |