Learn More
Kinesin 5B Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32279AF647
Product Code. 18730863
  • € 633.00 / 0.10mL
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
Alexa Fluor 647
Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain
The immunogen this antibody was made to, maps to a region between residue 913 to 963 of human Kinesin Family Member 5B using the numbering given in entry NP_004512.1 (GeneID 3799).
Primary
IgG
Polyclonal
50mM Sodium Borate
Rabbit
Immunogen affinity purified