missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5B Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-580AF750
This item is not returnable.
View return policy
Description
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Kinesin 5B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 750 | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 3799 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction