Learn More
Invitrogen™ Kinesin 5B Recombinant Rabbit Monoclonal Antibody (23GB4030), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553674
 View more versions of this product
Product Code. 30089325
  • € 562.00 / 100µL
Description
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kinesin 5B
Recombinant Monoclonal
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P33176, Q2PQA9, Q61768
KIF5B
A synthesized peptide derived from human KIF5B (10-45AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB4030
Unconjugated
KIF5B
AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745)
Rabbit
Affinity chromatography
RUO
117550, 16573, 3799
-20°C