missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen Antibody (MM0437-11L36), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11757JF646
This item is not returnable.
View return policy
Description
Kininogen Monoclonal antibody specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kininogen | |
| Monoclonal | |
| Janelia Fluor 646 | |
| BDK;BK;HAE6;HK;HMWK;Kininogen-1;KNG;KNG1 | |
| Human recombinant Kininogen | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 3827 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0437-11L36 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction