missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00 - € 470.00
Specifications
| Antigen | Kir2.1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383216
|
Novus Biologicals
NBP3-03675-20UL |
20 μL |
€ 193.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18304806
|
Novus Biologicals
NBP3-03675-100UL |
100 μL |
€ 470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kir2.1 Polyclonal specifically detects Kir2.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Kir2.1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3759 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Cardiac inward rectifier potassium channel, HHBIRK1, HHIRK1, hIRK1, Inward rectifier K(+) channel Kir2.1, inward rectifier K+ channel KIR2.1, inward rectifier potassium channel 2, IRK-1, IRK1LQT7, Kir2.1, Potassium channel, inwardly rectifying subfamily J member 2, potassium inwardly-rectifying channel, subfamily J, member 2, SQT3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 318-427 of human Kir2.1 (NP_000882.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title