missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kir2.2 Polyclonal antibody specifically detects Kir2.2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Kir2.2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | ATP-sensitive inward rectifier potassium channel 12, hIRK, hIRK1, hkir2.2x, Inward rectifier K(+) channel Kir2.2, Inward rectifier K(+) channel Kir2.2v, inward rectifier K(+) channel Kir2.6, IRK2IRK-2, kcnj12x, KCNJN1FLJ14167, Kir2.2, Kir2.2v, Potassium channel, inwardly rectifying subfamily J member 12, potassium inwardly-rectifying channel, subfamily J, inhibitor 1, potassium inwardly-rectifying channel, subfamily J, member 12 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 354-433 of human Kir2.2 (NP_066292.2).,, Sequence:, TPRCSAKDLVENKFLLPSANSFCYENELAFLSRDEEDEADGDQDGRSRDGLSPQARHDFDRLQAGGGVLEQRPYRRESEI |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?