missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL1/CD158a Antibody (NKVFS1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63267AF532
This item is not returnable.
View return policy
Description
KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| KIR2DL1/CD158a | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KIR2DL1 | |
| Made to Human KIR | |
| 0.1 mL | |
| Primary | |
| NB100-63267 recognizes KIR2D members of the killer cell immunoglobulin (Ig)-like receptor (KIR) family, CD158a, CD158b and P50.3. KIR2D family members are cell surface glycoproteins with two Ig domains, which are expressed on natural killer cells and some T cells. Clone NKVFS1 specifically recognizes the long and short forms CD158a and CD158b (KIR2DL, KIR2DS1 and KIR2DS2 respectively) and also p50.3 (KIR2DS4). The clone is reported to have functional activity, activating NK cell cytotoxicity via KIR2DS and inhibiting via KIR2DL forms. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA | |
| NKVFS1 | |
| Western Blot, Flow Cytometry, ELISA | |
| CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
| Mouse | |
| Protein A or G purified | |
| Immunology, Innate Immunity, Signal Transduction | |
| 3802.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction