missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL1/KIR2DS5 Antibody (143211), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1844MFV500
This item is not returnable.
View return policy
Description
KIR2DL1/KIR2DS5 Monoclonal antibody specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry, CyTOF-reported
Specifications
| KIR2DL1/KIR2DS5 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 143211 | |
| Flow Cytometry, CyTOF-reported | |
| CD158a | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5, Accession # P43626 | |
| RUO | |
| 3802 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction