Learn More
KIR2DL3/CD158b2 Antibody (180701), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2014PECY55
Product Code. 30026221
  • € 1230.00 / 0.50mL
Description
KIR2DL3/CD158b2 Monoclonal antibody specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIR2DL3/CD158b2
Monoclonal
PE-Cyanine5.5
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
180701
Flow Cytometry
CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2, Accession # P43628
0.5 mL
Apoptosis
3804
Store at 4C in the dark. Do not freeze.