missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIR2DS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525667
This item is not returnable.
View return policy
Description
KIR2DS3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
Specifications
| KIR2DS3 | |
| Polyclonal | |
| Unconjugated | |
| KIR2DS3 | |
| killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3; killer cell immunoglobulin-like receptor 2DS3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 3; KIR2DS3; MHC class I NK cell receptor; natural killer cell inhibitory receptor; natural killer-associated transcript 7; NKAT7; NKAT-7 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 3808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q14952 | |
| KIR2DS3 | |
| KLH conjugated synthetic peptide between 268-295 amino acids from the C-terminal region of human KIR2DS3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction