missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DS4/CD158i Antibody (MM0442-2G42), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-11762CL1
This item is not returnable.
View return policy
Description
KIR2DS4/CD158i Monoclonal antibody specifically detects KIR2DS4/CD158i in Human samples. It is validated for Flow Cytometry,Western Blot
Specifications
| KIR2DS4/CD158i | |
| Monoclonal | |
| CoraFluor 1 | |
| CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17 | |
| Human recombinant KIR2DS4/CD158i | |
| 0.1 mL | |
| Immunology | |
| 3809 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Flow Cytometry, Western Blot | |
| MM0442-2G42 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction