missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kir3.3 (KCNJ9) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15999774
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999774 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15999774 Supplier Invitrogen™ Supplier No. PA567106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RLDAHLYWSIP SRLDEKVEEE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir3.3 (KCNJ9)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNJ9
Gene Accession No. Q92806
Gene Alias 1700085N21Rik; G protein-activated inward rectifier potassium channel 3; G protein-coupled inward rectifier potassium channel; Girk3; GIRK-3; Inward rectifier K(+) channel Kir3.3; inwardly rectifier K(+) channel Kir3.3; inwardly rectifier K+ channel KIR3.3; KCNJ9; Kir3.3; mbGIRK3; Potassium channel, inwardly rectifying subfamily J member 9; potassium channel, inwardly rectifying subfamily J, member 9; potassium inwardly rectifying channel subfamily J member 9; potassium inwardly-rectifying channel subfamily J9; potassium inwardly-rectifying channel, subfamily J, member 9; potassium voltage-gated channel subfamily J member 9; si:dkey-42d18.2
Gene Symbols KCNJ9
Host Species Rabbit
Immunogen Recombinant Human KCNJ9. Recombinant protein control fragment (Product #RP-107750).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.