Learn More
KIR3DL1 Antibody (177407), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB12251I
Product Code. 18055725
  • ONLINE EXCLUSIVE PRICE
    € 915.00
    € 865.20 / 0.50mL
    Save € 49.80
    5% Off
Description
KIR3DL1 Monoclonal antibody specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIR3DL1
Monoclonal
Janelia Fluor 549
AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, NKAT-3, NKAT3KIR, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11
BaF3 mouse pro-B cell line transfected with human KIR3DL1, Accession # P43629
0.5 mL
Immunology
3811
Store at 4°C in the dark.
IgG2a
Flow Cytometry
177407
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant