missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DX1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00090011-B01P
This item is not returnable.
View return policy
Description
KIR3DX1 Polyclonal antibody specifically detects KIR3DX1 in Human samples. It is validated for Western Blot
Specifications
| KIR3DX1 | |
| Polyclonal | |
| Western Blot 1:500 | |
| FLJ00060, killer cell immunoglobulin-like receptor, three domains, X1, KIR3DL0, LENG12 | |
| KIR3DX1 (AAH33195.1, 1 a.a. - 78 a.a.) full-length human protein. MANTEPTEGQRTDEEEPAAEETQEIIYAQLNHQALSQTGFPPASQCPHYLSEDPSIYITVHQAQAEARAAPSLWHKGH | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 90011 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction