missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIR3DX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584101
This item is not returnable.
View return policy
Description
Immunogen sequence: CVGIYKHASK WSAESNSLKI IVTGLFTKPS ISAHPSSLVH AGARVSLRCH SELAFDEFIL YKEGHIQHSQ QLDQGME Highest antigen sequence indentity to the following orthologs: Rat - 42%, Mouse - 43%.
KIR3DX1 is a protein coding gene. Gene ontology (GO) annotation include extracellular region/cellular component.
Specifications
| KIR3DX1 | |
| Polyclonal | |
| Unconjugated | |
| KIR3DX1 | |
| FLJ00060; killer cell immunoglobulin like receptor, three Ig domains X1; KIR3DL0; KIR3DX1; LENG12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7L2 | |
| KIR3DX1 | |
| Recombinant protein corresponding to Human KIR3DX1. Recombinant protein control fragment (Product #RP-98913). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction