missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kir4.2 (KCNJ15) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15751411 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15751411 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15751411 Supplier Invitrogen™ Supplier No. PA553474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SAVCQSRTSY IPEEIYWGFE FVPVVSLSKN GKYVADFSQF EQIRKSPDCT FYCADSEKQQ LEEKYRQEDQ RERELRTL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir4.2 (KCNJ15)
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnj15
Gene Accession No. Q99712
Gene Alias 4930414N08Rik; AI182284; AI267127; ATP-sensitive inward rectifier potassium channel 15; Inward rectifier K(+) channel Kir1.3; inward rectifier K(+) channel Kir4.2; inward rectifier K+ channel KIR4.2; IRKK; KCNJ14; KCNJ15; KIR1.3; Kir4.2; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; potassium voltage-gated channel subfamily J member 15
Gene Symbols Kcnj15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNJ15. Recombinant protein control fragment (Product #RP-91000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3772
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.