missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kirrel2/NEPH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68977
This item is not returnable.
View return policy
Description
Kirrel2/NEPH3 Polyclonal antibody specifically detects Kirrel2/NEPH3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Kirrel2/NEPH3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| DKFZp564A1164, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1kin of irregular chiasm-like 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EVTLSASPHTVQEGEKVIFLCQATAQPPVTGYRWAKGGSPVLGARGPRLEVVADASFL | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84063 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction