Learn More
Invitrogen™ KISS1R Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA596221
Product Code. 16549061
  • € 528.00 / 100µL
Description
Positive Samples: SW480; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: ALHRLSPGPR AY.
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KISS1R
Polyclonal
Unconjugated
KISS1R
AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175
Rabbit
Affinity Chromatography
RUO
114229, 78976, 84634
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.31 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Q91V45, Q924U1, Q969F8
KISS1R
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human KISS1R (NP_1159402).
100 μL
Primary
Human, Mouse, Rat