missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIST Antibody (OTI2H4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72383PE
This item is not returnable.
View return policy
Description
KIST Monoclonal specifically detects KIST in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| KIST | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| UHMK1 | |
| Full length recombinant protein of human KIST (NP_787062) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI2H4 | |
| Flow Cytometry | |
| DKFZp434C1613, EC 2.7.11.1, FLJ23015, kinase interacting with leukemia-associated gene (stathmin), Kinase interacting with stathmin, KISKIS protein kinase, KIST, serine/threonine-protein kinase Kist, U2AF homology motif (UHM) kinase 1, U2AF homology motif kinase 1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 127933.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction