missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIT, Mouse anti-Human, Clone: 2C3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003815-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: PGKSDLIVRVGDEIRLLCTDPGFVKWTFEILDETNENKQNEWITEKAEATNTGKYTCTNKHGLSNSIYVFVRDPAKLFLVDRSLYGKEDNDTLVRCPLTDSpecifications
| KIT | |
| Monoclonal | |
| Unconjugated | |
| v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog | |
| C-Kit/CD117/PBT/SCFR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2C3 | |
| In 1x PBS, pH 7.4 | |
| NM_000222 | |
| KIT | |
| KIT (NP_000213, 41 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3815 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction