missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KJ Pyr 9

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
KJ Pyr 9 is a high affinity Myc inhibitor (Kd = 6.5 nM). Disrupts Myc-Max interaction. Inhibits Myc-dependent cancer cell line proliferation in vitro. Inhibits growth of MDA-MB-231 breast cancer cell xenografts in mice. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 4-[2-(2-Furanyl)-6-(4-nitrophenyl)-4-pyridinyl]benzamide |
| CAS | 581073-80-5 |
| Quantity | 10 mg |
| Target | Additional Transcription Factor Compounds |
| Molecular Formula | C22H15N3O4 |
| Formula Weight | 385.37 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction