missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | KLC1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227453
|
Novus Biologicals
NBP3-38165-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232186
|
Novus Biologicals
NBP3-38165-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLC1 Polyclonal antibody specifically detects KLC1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| KLC1 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 3831 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hKLC1B, hKLC1G, hKLC1J, hKLC1S, kinesin 2, kinesin 2 60/70kDa, kinesin light chain 1, KLC 1, KLChKLC1N, KNS2AhKLC1P, KNS2hKLC1R, medulloblastoma antigen MU-MB-2.50, MGC15245 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 261-560 of human KLC1 (KLC1 (KLC1)) (NP_005543.2).,, Sequence:, LALVYRDQNKYKDAANLLNDALAIREKTLGKDHPAVAATLNNLAVLYGKRGKYKEAEPLCKRALEIREKVLGKDHPDVAKQLNNLALLCQNQGKYEEVEYYYQRALEIYQTKLGPDDPNVAKTKNNLASCYLKQGKFKQAETLYKEILTRAHEREFGSVDDENKPIWMHAEEREECKGKQKDGTSFGEYGGWYKACKVDSPTVTTTLKNLGALYRRQGKFEAAETLEEAAMRSRKQGLDNVHKQRVAEVLNDPENMEKRRSRESLNVDVVKYESGPDGGEEVSMSVEWNGMRKMKLGLVN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title