missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562613
This item is not returnable.
View return policy
Description
Immunogen sequence: EEAAMRSRKQ GLDNVHKQRV AEVLNDPENM EKRRSRESLN VDVVKYESGP DGGEEVSMSV EWNG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Conventional kinesin is a tetrameric molecule composed of two heavy chains and two light chains, and transports various cargos along microtubules toward their plus ends. The heavy chains provide the motor activity, while the light chains bind to various cargos. This gene encodes a member of the kinesin light chain family. It associates with kinesin heavy chain through an N-terminal domain, and six tetratricopeptide repeat (TPR) motifs are thought to be involved in binding of cargos such as vesicles, mitochondria, and the Golgi complex. Thus, kinesin light chains function as adapter molecules and not motors per se. Although previously named 'kinesin 2', this gene is not a member of the kinesin-2 / kinesin heavy chain subfamily of kinesin motor proteins. Extensive alternative splicing produces isoforms with different C-termini that are proposed to bind to different cargos; however, the full-length nature of some of these variants has not been determined.
Specifications
| KLC1 | |
| Polyclonal | |
| Unconjugated | |
| KLC1 | |
| AI874768; kinesin 2; kinesin 2 60/70kDa; kinesin light chain 1; Kinesin2; Klc; KLC 1; Klc1; KNS 2; KNS 2A; Kns2; KNS2A; medulloblastoma antigen MU-MB-2.50; MGC15245 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q07866 | |
| KLC1 | |
| Recombinant protein corresponding to Human KLC1. Recombinant protein control fragment (Product #RP-97607). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction