missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLC4 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06417JF549
This item is not returnable.
View return policy
Description
KLC4 Polyclonal antibody specifically detects KLC4 in Human samples. It is validated for Western Blot
Specifications
| KLC4 | |
| Polyclonal | |
| Western Blot | |
| bA387M24.3, kinesin light chain 4, kinesin-like 8, Kinesin-like protein 8, KLC 4, KNSL8, MGC111777 | |
| Produced in rabbits immunized with E. coli-derived Human KLC4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 89953 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction