Learn More
Ecolab™ Klercide Sporicidal Active Chlorine
Product Code. 15352428
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15352428 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15352428 Supplier Ecolab™ Supplier No. 3078930
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Deisgned to control all types of microbial contamination in cleanrooms. Ecolab™ Klercide Sporicidal Active Chloring is an effective biocide based on active chlorine, available in a ready to use spray bottle.
  • Sterile by gamma irradiation
  • EN 1276 Bactericidal; EN 1650 Fungicidal (yeast); EN 13697 Bactericidal and fungicidal
  • Cleanroom processed
  • Unique Sterishield Delivery System ensures the sterility of the contents is preserved throughout use.
  • Available with complete supporting documentation
Specifications
Product Type Biocide
Sterility Sterile
Appearance Colorless
Certifications/Compliance Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and fungicidal EN 12397; Sporicidal EN 13704; Virucidal EN 14476
Container Type Spray Bottle
Description Klercide sporicidal active chlorine; Sporicidal Active Chlorine
For Use With (Application) Bactericidal, fungicidal, sporicidal, virucidal agent
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • Klercide Sporicidal Active Chlorine
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: sodium hypochlorite