missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF1 Antibody (1B6A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 228.00 - € 578.00
Specifications
| Antigen | KLF1 |
|---|---|
| Clone | 1B6A3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693616
|
Novus Biologicals
NBP2-37380-0.025ml |
0.025 mL |
€ 228.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18195509
|
Novus Biologicals
NBP2-37380 |
0.1 mL |
€ 578.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF1 Monoclonal specifically detects KLF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| KLF1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| EKLFerythroid-specific transcription factor EKLF, Erythroid krueppel-like transcription factor, erythroid Kruppel-like factor, HBFQTL6, INLU, Krueppel-like factor 1, Kruppel-like factor 1 (erythroid), monoclonal A3D8 | |
| KLF1 | |
| IgG1 | |
| Affinity Purified |
| 1B6A3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 10661 | |
| Purified recombinant fragment of human KLF1 (AA: 208-362) expressed in E. Coli. | |
| Primary | |
| 38.2 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title