missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300c Antibody (072), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89991F
This item is not returnable.
View return policy
Description
CD300c Monoclonal specifically detects CD300c in Human samples. It is validated for ELISA.
Specifications
| CD300c | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 72 | |
| ELISA | |
| CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300c (NP_006669.1; Met1-Arg183). | |
| 0.1 mL | |
| Immunology | |
| 10871 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction