missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF11 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810999
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| KLF11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14901 | |
| KLF11 | |
| Human recombit protein fragment corresponding to amino acids 1-274 of human KLF11 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G4 | |
| Unconjugated | |
| KLF11 | |
| FKLF, FKLF1, MODY7, TIEG2, Tieg3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8462 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction