Learn More
KLF12 Antibody (1230C) [PE], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9097P
This item is not returnable.
View return policy
Description
KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| KLF12 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1230C | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
| Mouse myeloma cell line NS0-derived human KLF12, Asn2-Val402, Accession # Q9Y4X4 | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 11278 | |
| Store at 4°C in the dark. | |
| IgG |