missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Monoclonal Antibody (OTI3D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810315
This item is not returnable.
View return policy
Description
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| KLF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y4X4 | |
| KLF12 | |
| Full length human recombit protein of human KLF12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3D4 | |
| Unconjugated | |
| KLF12 | |
| AP-2rep, AP2REP, HSPC122 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11278 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction