missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF17 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82001F
This item is not returnable.
View return policy
Description
KLF17 Polyclonal antibody specifically detects KLF17 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| KLF17 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 128209 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| FLJ40160, Kruppel-like factor 17, Zfp393, Zinc finger protein 393Krueppel-like factor 17, ZNF393novel zinc-finger protein | |
| KLF17 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human KLF17. The immunogen is located within the last 50 amino acids of KLF17. Amino Acid Squence: SREFMRSDHLKQHQKTH | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction