missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF2 Antibody (665333), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5466CL1
This item is not returnable.
View return policy
Description
KLF2 Monoclonal antibody specifically detects KLF2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| KLF2 | |
| Monoclonal | |
| CoraFluor 1 | |
| Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
| E. coli-derived recombinant human KLF2, Pro71-Pro168 (predicted), Accession # Q9Y5W3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 665333 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10365 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction