missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF2 Antibody (OTI3A10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71597C
This item is not returnable.
View return policy
Description
KLF2 Monoclonal specifically detects KLF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| KLF2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KLF2 | |
| Full length recombinant protein of human KLF2 (NP_057354) produced in E.coli. | |
| 0.1 mL | |
| 10365.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3A10 | |
| Western Blot, Immunohistochemistry | |
| Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction