missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32245
This item is not returnable.
View return policy
Description
KLF5 Polyclonal specifically detects KLF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| KLF5 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH7), 20% Glycerol with 0.025% Proclin 300 | |
| KLF5 | |
| Recombinant protein encompassing a sequence within the center region of human KLF5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen | |
| Basic transcription element-binding protein 2, BTEB2, BTE-binding protein 2, CKLFGC-box-binding protein 2, Colon krueppel-like factor, colon kruppel-like factor, GC box binding protein 2, IKLFbasic transcription element binding protein 2, Intestinal-enriched krueppel-like factor, intestinal-enriched kruppel-like factor, Krueppel-like factor 5, Kruppel-like factor 5 (intestinal), Transcription factor BTEB2 | |
| Rabbit | |
| 51 kDa | |
| 100 μL | |
| Cancer | |
| 688 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction