Learn More
KLF5 Mouse anti-Human, Clone: OTI2G4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811879
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.Specifications
| KLF5 | |
| Monoclonal | |
| Unconjugated | |
| KLF5 | |
| BTEB2, CKLF, IKLF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 688 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13887 | |
| KLF5 | |
| Human recombit protein fragment corresponding to amino acids 1-457 of human KLF5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.