missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KLF8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16374345 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16374345 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16374345 Supplier Invitrogen™ Supplier No. PA595077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SMMC Cell, 293T Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF8
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene KLF8
Gene Accession No. O95600
Gene Alias A830097P10Rik; Basic krueppel-like factor 3; basic kruppel-like factor 3; BKLF3; DKFZp686O08126; DXS741; KLF8; Klf8-ps1; Krueppel-like factor 8; Kruppel like factor 8; Kruppel-like factor 8; Kruppel-like factor 8, pseudogene 1; LOW QUALITY PROTEIN: Krueppel-like factor 8; MGC138314; RGD1565831; RP13-1021K9.1; zinc finger protein 741; ZNF74; ZNF741
Gene Symbols KLF8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human KLF8 (1-18aa MVDMDKLINNLEVQLNSE).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11279
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.