missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF9 Antibody (OTI8A11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70285AF700
This item is not returnable.
View return policy
Description
KLF9 Monoclonal specifically detects KLF9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| KLF9 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KLF9 | |
| Full length recombinant protein of human KLF9 (NP_001197) produced in E.coli. | |
| 0.1 mL | |
| 687.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI8A11 | |
| Western Blot, Immunohistochemistry | |
| basic transcription element binding protein 1, Basic transcription element-binding protein 1, BTEB1GC-box-binding protein 1, BTEBBTE-binding protein 1, Krueppel-like factor 9, Kruppel-like factor 9, Transcription factor BTEB1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction