missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF9 Monoclonal Antibody (OTI7H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808445S
This item is not returnable.
View return policy
Description
KLF9 Monoclonal antibody specifically detects KLF9 in Human samples. It is validated for Western BlotSpecifications
| KLF9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13886 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI7H8 | |
| Unconjugated | |
| KLF9 | |
| 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1 | |
| Full length human recombinant protein of human KLF9 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction