missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF9 Monoclonal Antibody (OTI7H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808445
This item is not returnable.
View return policy
Description
KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregcy and tumorigenesis.Specifications
| KLF9 | |
| Monoclonal | |
| Unconjugated | |
| KLF9 | |
| BTEB, BTEB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7H8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13886 | |
| KLF9 | |
| Full length human recombit protein of human KLF9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction