missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH Antibody, Alexa Fluor™ 750 , Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-78810AF750
This item is not returnable.
View return policy
Description
KLH Polyclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| KLH | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Alexa Fluor 750 | |
| keyhole limpet hemocyanin (KLH) | |
| KLH was used as the immunogen. | |
| 1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur