missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554215
This item is not returnable.
View return policy
Description
Immunogen sequence: FLCGGLSADN IPLSDGWIHN VTTNCWKQLT HLPKTRPRLW HTACLGKENE IMVFGGSKDD LLALDTGHCN DLLIFQTQPY SLLRSCLDCI GKNSIMLESQ ISLLPPKLLQ QVLKKITFWA AANHREEQRV Q Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.
Specifications
| KLHDC1 | |
| Polyclonal | |
| Unconjugated | |
| KLHDC1 | |
| kelch domain containing 1; kelch domain-containing protein 1; Klhdc1; MST025; MSTP025 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 122773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7A1 | |
| KLHDC1 | |
| Recombinant protein corresponding to Human KLHDC1. Recombinant protein control fragment (Product #RP-88913). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction