missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHDC5 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18445JF585
This item is not returnable.
View return policy
Description
KLHDC5 Polyclonal antibody specifically detects KLHDC5 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| KLHDC5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57542 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| Ctb9, kelch domain containing 5, kelch domain-containing protein 5, KIAA1340, MGC131714 | |
| Recognizes region between residues Between Between 400 and 450.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction