missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17076
This item is not returnable.
View return policy
Description
KLHL1 Polyclonal specifically detects KLHL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| KLHL1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| KLHL1 | |
| Recombinant protein encompassing a sequence within the center region of human KLHL1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 57626 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.03mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FLJ30047, kelch (Drosophila)-like 1, kelch-like 1 (Drosophila), KIAA1490kelch-like protein 1, Mayven-related protein 2, MRP2 | |
| Rabbit | |
| 83 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Monkey (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction