missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL12/C3IP1 Antibody (OTI4E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72118CL1
This item is not returnable.
View return policy
Description
KLHL12/C3IP1 Monoclonal antibody specifically detects KLHL12/C3IP1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| KLHL12/C3IP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
| Human recombinant protein fragment corresponding to amino acids 177-405 of human KLHL12 (NP_067646) produced in E.coli. | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 59349 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI4E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction