missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL12/C3IP1 Antibody (OTI4E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45512
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
KLHL12/C3IP1 Monoclonal specifically detects KLHL12/C3IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| KLHL12/C3IP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
| Mouse | |
| 63.1 kDa | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 59349 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI4E1 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| KLHL12 | |
| Human recombinant protein fragment corresponding to amino acids 177-405 of human KLHL12 (NP_067646) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction