missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLHL12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567166
This item is not returnable.
View return policy
Description
Immunogen sequence: FAETHNCVDLM QAAEVFSQKH FPEVVQHEEF ILLSQGEVEK LIKCDEIQVD SEEPV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
Specifications
| KLHL12 | |
| Polyclonal | |
| Unconjugated | |
| Klhl12 | |
| C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 59349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53G59 | |
| Klhl12 | |
| Recombinant Human KLHL12. Recombinant protein control fragment (Product #RP-105530). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction