missing translation for 'onlineSavingsMsg'
Learn More

KLHL2 Antibody (OTI1G7), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71695JF646

Product Code. 18192894

  • ONLINE EXCLUSIVE PRICE
    € 785.00
    € 742.35 / 0.10mL
    Save € 42.65
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KLHL2 Monoclonal specifically detects KLHL2 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KLHL2
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
KLHL2
Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells.
0.1 mL
11275.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1G7
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Canine, Monkey, Primate
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.